Assay Details
| Gene Symbol: Maf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164608.1 | NP_001158080.1 | 1 - 2 | 366 | 72 | |
| GenBank mRNA | AK042913.1 | - | 1 - 2 | 307 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042913.1, NM_001164608.1 transcripts.
Set Membership: |
|

Maf1
Genomic Map