Assay Details
| Gene Symbol: Gpihbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026730.1 | NP_081006.1 | 1 - 2 | 75 | 68 | |
| GenBank mRNA | AB095543.1 | - | 1 - 2 | 82 | 68 | |
| AK003305.1 | - | 1 - 2 | 91 | 68 | ||
| AK050431.1 | - | 80 | 68 | |||
| BC061225.1 | - | 1 - 2 | 75 | 30298145 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpihbp1
Genomic Map