Assay Details
| Gene Symbol: Gpihbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026730.1 | NP_081006.1 | 2 - 3 | 193 | 74 | |
| GenBank mRNA | AB095543.1 | - | 2 - 3 | 200 | 74 | |
| AK003305.1 | - | 2 - 3 | 209 | 74 | ||
| AK050431.1 | - | 198 | 74 | |||
| BC061225.1 | - | 2 - 3 | 193 | 30298145 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpihbp1
Genomic Map