Assay Details
| Gene Symbol: Oc90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010953.2 | NP_035083.1 | 2 - 3 | 145 | 85 | |
| GenBank mRNA | AF093591.1 | - | 2 - 3 | 255 | 85 | |
| AK012981.1 | - | 2 - 3 | 147 | 85 | ||
| BC048716.1 | - | 2 - 3 | 112 | 6334417 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oc90
Genomic Map