Assay Details
| Gene Symbol: Oc90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010953.2 | NP_035083.1 | 3 - 4 | 205 | 78 | |
| GenBank mRNA | AF091846.1 | - | 1 - 2 | 59 | 78 | |
| AF093591.1 | - | 3 - 4 | 315 | 78 | ||
| AK012981.1 | - | 3 - 4 | 207 | 78 | ||
| BC048716.1 | - | 3 - 4 | 172 | 6334417 | 78 | |
More Information
Set Membership: |
|

Oc90
Genomic Map