Assay Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175151.4 | NP_780360.1 | 2 - 3 | 113 | 103 | |
| GenBank mRNA | AK042248.1 | - | 2 - 3 | 114 | 103 | |
| AK161972.1 | - | 2 - 3 | 122 | 103 | ||
| BC061187.1 | - | 2 - 3 | 110 | 30277245 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tatdn1
Genomic Map