Assay Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175151.4 | NP_780360.1 | 8 - 9 | 546 | 213 | |
| GenBank mRNA | AK042248.1 | - | 8 - 9 | 547 | 213 | |
| AK161972.1 | - | 8 - 9 | 631 | 213 | ||
| BC034895.1 | - | 5 - 6 | 333 | 127 | ||
| BC061187.1 | - | 8 - 9 | 543 | 30277245 | 213 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tatdn1
Genomic Map