Assay Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028364.2 | NP_082640.2 | 5 - 6 | 438 | 88 | |
| GenBank mRNA | AK146534.1 | - | 5 - 6 | 408 | 88 | |
| AK149632.1 | - | 5 - 6 | 404 | 88 | ||
| BC119437.1 | - | 2 - 3 | 94 | 88 | ||
| BC148274.1 | - | 2 - 3 | 94 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Puf60
Genomic Map