Assay Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030565.6 | NP_085042.2 | 6 - 7 | 1788 | 62 | |
| GenBank mRNA | AK149780.1 | - | 7 - 8 | 1868 | 62 | |
| AK154810.1 | - | 6 - 7 | 1784 | 62 | ||
| AK220286.1 | - | 6 - 7 | 1304 | 62 | ||
| AY778962.1 | - | 6 - 7 | 1769 | 62 | ||
| BC004044.1 | - | 3 - 4 | 397 | 62 | ||
| BC025814.1 | - | 6 - 7 | 1739 | 62 | ||
| BC025826.1 | - | 6 - 7 | 1769 | 62 | ||
| CT010363.1 | - | 2 - 3 | 243 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam20c
Genomic Map