Assay Details
| Gene Symbol: Stambpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029682.4 | NP_083958.3 | 5 - 6 | 836 | 121 | |
| GenBank mRNA | AB010121.1 | - | 5 - 6 | 531 | 121 | |
| AB010122.1 | - | 5 - 6 | 495 | 121 | ||
| AB066211.1 | - | 5 - 6 | 836 | 121 | ||
| AB066213.1 | - | 5 - 6 | 519 | 121 | ||
| AK085665.1 | - | 4 - 5 | 3227 | 121 | ||
| BC018343.1 | - | 5 - 6 | 562 | 4010423 | 121 | |
| BC023097.1 | - | 5 - 6 | 805 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stambpl1
Genomic Map