Assay Details
| Gene Symbol: Srrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021403.3 | NP_067378.2 | 8 - 9 | 1198 | 80 | |
| GenBank mRNA | BC051947.1 | - | 8 - 9 | 805 | 5696610 | 80 |
| BC058385.1 | - | 8 - 9 | 1198 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm3
Genomic Map