Assay Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159571.1 | NP_001153043.1 | 10 - 11 | 2271 | 85 | |
| NM_010144.6 | NP_034274.4 | 10 - 11 | 2244 | 85 | ||
| GenBank mRNA | AK046884.1 | - | 10 - 11 | 2209 | 85 | |
| AK049340.1 | - | 10 - 11 | 2245 | 85 | ||
| AK132392.1 | - | 10 - 11 | 2244 | 85 | ||
| AK143422.1 | - | 2214 | 85 | |||
| BC013342.1 | - | 10 - 11 | 1932 | 85 | ||
| BC016505.1 | - | 10 - 11 | 2196 | 3709926 | 85 | |
| BC090839.1 | - | 10 - 11 | 2021 | 85 | ||
| U06834.1 | - | 11 - 12 | 1857 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb4
Genomic Map