Assay Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010144.6 | NP_034274.4 | 6 - 7 | 1786 | 61 | |
| GenBank mRNA | AK049340.1 | - | 6 - 7 | 1787 | 61 | |
| AK132392.1 | - | 6 - 7 | 1786 | 61 | ||
| AK143422.1 | - | 1756 | 61 | |||
| BC016505.1 | - | 6 - 7 | 1738 | 3709926 | 61 | |
| BC090839.1 | - | 6 - 7 | 1563 | 61 | ||
| U06834.1 | - | 7 - 8 | 1399 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb4
Genomic Map