Assay Details
| Gene Symbol: Clip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039162.2 | NP_001034251.1 | 7 - 8 | 1776 | 104 | |
| NM_009990.3 | NP_034120.2 | 7 - 8 | 1776 | 104 | ||
| GenBank mRNA | AK045967.1 | - | 7 - 8 | 1620 | 104 | |
| AK172928.1 | - | 7 - 8 | 1521 | 104 | ||
| BC039162.1 | - | 7 - 8 | 1716 | 3498067 | 104 | |
| BC053048.1 | - | 7 - 8 | 1776 | 5709279 | 104 | |
| BC121191.1 | - | 7 - 8 | 1422 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clip2
Genomic Map