Assay Details
| Gene Symbol: Scarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205082.1 | NP_001192011.1 | 11 - 12 | 1613 | 89 | |
| GenBank mRNA | AK010622.1 | - | 11 - 12 | 1538 | 89 | |
| AK171602.1 | - | 11 - 12 | 1593 | 89 | ||
| AK187730.1 | - | 1 - 2 | 115 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scarb1
Genomic Map