Assay Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008617.2 | NP_032643.2 | 8 - 9 | 974 | 62 | |
| GenBank mRNA | AK002305.1 | - | 8 - 9 | 991 | 62 | |
| AK135162.1 | - | 8 - 9 | 987 | 62 | ||
| AK160553.1 | - | 8 - 9 | 957 | 62 | ||
| AK167809.1 | - | 8 - 9 | 957 | 62 | ||
| BC023482.1 | - | 8 - 9 | 974 | 5251192 | 62 | |
| DQ402950.1 | - | 7 - 8 | 824 | 62 | ||
| M16229.1 | - | 8 - 9 | 933 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
tricarboxylic acid cycle
oxaloacetate metabolic process
malate metabolic process
internal protein amino acid acetylation
NADH metabolic process
carboxylic acid metabolic process
oxidation-reduction process
catalytic activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
L-malate dehydrogenase activity
protein self-association
poly(A) RNA binding
malate dehydrogenase (NADP+) activity
carbohydrate metabolic process
tricarboxylic acid cycle
oxaloacetate metabolic process
malate metabolic process
internal protein amino acid acetylation
NADH metabolic process
carboxylic acid metabolic process
oxidation-reduction process
catalytic activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
L-malate dehydrogenase activity
protein self-association
poly(A) RNA binding
malate dehydrogenase (NADP+) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117834 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864719 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817622 | AK028047.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817622 | BC006844.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817622 | BC031768.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817622 | NM_177614.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48482 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168385 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151425 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | M16229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | M16229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117857 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168385 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01229107 | - | - |
| Whole Mouse Genome G4122A | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | M16229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK135162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK160553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | AK167809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384879 | BC023482.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136845 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168385 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04807449 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04807434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04807435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04729239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04703268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04807435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04321740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04321739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04194331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04884785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04271276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04807443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04496684 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526389 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416478_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433984_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526381 | - | - |
| Mouse Genome 430 2.0 Array | 1416478_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416478_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1433984_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1433984_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17448_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17448_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500014312.mm.1 | TC0500001586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014312.mm.1 | NM_008617.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014312.mm.1 | NM_008617.2 | Refseq |
| Mouse Expression Set 430 | 1433984_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416478_PM_a_at | - | - |
| Mouse Expression Set 430 | 1433984_a_at | - | - |
| Mouse Expression Set 430 | 1416478_a_at | - | - |
| Murine Genome U74v2 Set | 93991_PM_at | - | - |
| Murine Genome U74v2 Set | 93991_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416478_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433984_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433984_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416478_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416478_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416478_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433984_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433984_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 342300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1850601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2766244 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2766244 | - | - |
| mouse-6 v1.1 (Illumina) | scl017448.9_233-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017448.9_233-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2873112 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2873112 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017448.9_233-S | - | - |

Mdh2
Genomic Map