Assay Details
| Gene Symbol: Slc15a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133895.1 | NP_598656.1 | 6 - 7 | 1435 | 68 | |
| GenBank mRNA | AK048529.1 | - | 6 - 7 | 1402 | 68 | |
| AK154238.1 | - | 6 - 7 | 1403 | 68 | ||
| AK179597.1 | - | 2 - 3 | 241 | 68 | ||
| BC016233.1 | - | 6 - 7 | 1435 | 4502184 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a4
Genomic Map