Assay Details
| Gene Symbol: Sparcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010097.4 | NP_034227.3 | 3 - 4 | 913 | 100 | |
| GenBank mRNA | AK134608.1 | - | 3 - 4 | 554 | 100 | |
| AK158725.1 | - | 3 - 4 | 420 | 100 | ||
| BC003759.1 | - | 3 - 4 | 558 | 3486162 | 100 | |
| U64827.1 | - | 3 - 4 | 511 | 100 | ||
| U77330.1 | - | 3 - 4 | 578 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sparcl1
Genomic Map