Assay Details
| Gene Symbol: Hnrnpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077265.2 | NP_001070733.1 | 3 - 4 | 772 | 144 | |
| NM_001077266.2 | NP_001070734.1 | 2 - 3 | 715 | 144 | ||
| NM_001077267.2 | NP_001070735.1 | 2 - 3 | 715 | 144 | ||
| NM_007516.3 | NP_031542.2 | 3 - 4 | 772 | 144 | ||
| GenBank mRNA | AK077409.1 | - | 3 - 4 | 761 | 144 | |
| AK135733.1 | - | 3 - 4 | 718 | 144 | ||
| BC011172.1 | - | 3 - 4 | 447 | 144 | ||
| BC049098.1 | - | 3 - 4 | 772 | 5717286 | 144 | |
| CK618758.1 | - | 2 - 3 | 341 | 144 | ||
| U11273.1 | - | 2 - 3 | 552 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpd
Genomic Map