Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700002146.mm.1 |
NM_145484.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002146.mm.1 |
NM_145484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002146.mm.1 |
TC1700000290.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002147.mm.1 |
NM_145484.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002147.mm.1 |
NM_145484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002147.mm.1 |
TC1700000290.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002145.mm.1 |
NM_145484.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002145.mm.1 |
NM_145484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002145.mm.1 |
TC1700000290.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002144.mm.1 |
NM_145484.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002144.mm.1 |
NM_145484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002144.mm.1 |
TC1700000290.mm.1 |
MTA TC |