Assay Details
| Gene Symbol: Tgfbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011578.3 | NP_035708.2 | 12 - 13 | 2291 | 139 | |
| GenBank mRNA | AF039601.1 | - | 12 - 13 | 2091 | 139 | |
| AK133234.1 | - | 10 - 11 | 1801 | 139 | ||
| BC070428.1 | - | 12 - 13 | 2216 | 6410554 | 139 | |
More Information
Set Membership: |
|

Tgfbr3
Genomic Map