Assay Details
| Gene Symbol: Tgfbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011578.3 | NP_035708.2 | 16 - 17 | 2869 | 103 | |
| GenBank mRNA | AF039601.1 | - | 16 - 17 | 2669 | 103 | |
| BC070428.1 | - | 16 - 17 | 2794 | 6410554 | 103 | |
More Information
Set Membership: |
|

Tgfbr3
Genomic Map