Assay Details
| Gene Symbol: Tgfbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011578.3 | NP_035708.2 | 9 - 10 | 1838 | 67 | |
| GenBank mRNA | AF039601.1 | - | 9 - 10 | 1638 | 67 | |
| AK133234.1 | - | 7 - 8 | 1348 | 67 | ||
| BC070428.1 | - | 9 - 10 | 1763 | 6410554 | 67 | |
More Information
Set Membership: |
|

Tgfbr3
Genomic Map