Assay Details
| Gene Symbol: Atp2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110140.3 | NP_001103610.1 | 2 - 3 | 670 | 81 | |
| NM_009722.3 | NP_033852.1 | 2 - 3 | 670 | 81 | ||
| NR_027838.1 | 2 - 3 | 670 | 81 | |||
| GenBank mRNA | AJ131821.1 | - | 2 - 3 | 340 | 81 | |
| AJ223584.1 | - | 2 - 3 | 340 | 81 | ||
| AK220538.1 | - | 2 - 3 | 619 | 81 | ||
| BC054531.1 | - | 2 - 3 | 329 | 6406916 | 81 | |
| BC054748.1 | - | 2 - 3 | 329 | 6406916 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp2a2
Genomic Map