Assay Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144915.3 | NP_659164.2 | 8 - 9 | 1324 | 80 | |
| GenBank mRNA | AK086747.1 | - | 8 - 9 | 1236 | 80 | |
| AK087884.1 | - | 8 - 9 | 1179 | 80 | ||
| AK136372.1 | - | 8 - 9 | 1182 | 80 | ||
| BC016105.1 | - | 8 - 9 | 1220 | 4921222 | 80 | |
More Information
Set Membership: |
|

Daglb
Genomic Map