Assay Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144915.3 | NP_659164.2 | 4 - 5 | 868 | 88 | |
| GenBank mRNA | AK086747.1 | - | 4 - 5 | 780 | 88 | |
| AK087884.1 | - | 4 - 5 | 723 | 88 | ||
| AK136372.1 | - | 4 - 5 | 726 | 88 | ||
| BC016105.1 | - | 4 - 5 | 764 | 4921222 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Daglb
Genomic Map