Assay Details
| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031084.1 | - | 1 - 2 | 78 | 73 | |
| AK149630.1 | - | 1 - 2 | 73 | 73 | ||
| BC005606.1 | - | 1 - 2 | 65 | 3498140 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149630.1, BC005606.1 transcripts.
Set Membership: |
|

Mvk
Genomic Map