Assay Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286396.1 | NP_001273325.1 | 3 - 4 | 355 | 71 | |
| NM_001286397.1 | NP_001273326.1 | 3 - 4 | 365 | 71 | ||
| NM_010018.3 | NP_034148.2 | 3 - 4 | 514 | 71 | ||
| GenBank mRNA | AK134813.1 | - | 3 - 4 | 375 | 71 | |
| BC015269.1 | - | 3 - 4 | 365 | 4239778 | 71 | |
| BC018377.1 | - | 3 - 4 | 355 | 4242375 | 71 | |
| D10210.1 | - | 2 - 3 | 306 | 71 | ||
| D10211.1 | - | 2 - 3 | 306 | 71 | ||
| M32299.1 | - | 3 - 4 | 371 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dao
Genomic Map