Assay Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286396.1 | NP_001273325.1 | 5 - 6 | 503 | 63 | |
| NM_001286397.1 | NP_001273326.1 | 5 - 6 | 513 | 63 | ||
| NM_010018.3 | NP_034148.2 | 5 - 6 | 662 | 63 | ||
| GenBank mRNA | AK134813.1 | - | 5 - 6 | 523 | 63 | |
| BC015269.1 | - | 5 - 6 | 513 | 4239778 | 63 | |
| BC018377.1 | - | 5 - 6 | 503 | 4242375 | 63 | |
| D10211.1 | - | 4 - 5 | 454 | 63 | ||
| M32299.1 | - | 5 - 6 | 519 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dao
Genomic Map