| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
TC0500002927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_001161738.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_001161738.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_001161739.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_001161739.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_133248.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025877.mm.1 |
NM_133248.2 |
Refseq |