Assay Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148927.2 | NP_683729.2 | 16 - 17 | 1874 | 74 | |
| GenBank mRNA | BC024961.1 | - | 15 - 16 | 1919 | 4945500 | 74 |
| BC030731.1 | - | 10 - 11 | 1078 | 74 | ||
| BE652135.1 | - | 4 - 5 | 381 | 74 | ||
More Information
Set Membership: |
|

Plekha4
Genomic Map