| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
TC0500001668.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256116.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256116.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256117.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256118.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_001256118.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_024451.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500015380.mm.1 |
NM_024451.2 |
Refseq |