Assay Details
| Gene Symbol: Lamtor4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081108.2 | NP_001074577.1 | 3 - 4 | 520 | 57 | |
| GenBank mRNA | AK002488.1 | - | 3 - 4 | 247 | 57 | |
| AK003086.1 | - | 3 - 4 | 271 | 57 | ||
| AK003250.1 | - | 3 - 4 | 230 | 57 | ||
| AK010598.1 | - | 3 - 4 | 252 | 57 | ||
| AK146994.1 | - | 3 - 4 | 228 | 57 | ||
| BC056178.1 | - | 2 - 3 | 217 | 57 | ||
| BC144958.1 | - | 3 - 4 | 520 | 9053506 | 57 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | BC056178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK146994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | NM_001081108.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK002488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK003086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | BC056178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK146994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | NM_001081108.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK002488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P316405 | AK003086.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746100 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127221 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | BC056178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK146994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | NM_001081108.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK002488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK003086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | BC056178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK146994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | NM_001081108.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK002488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P316405 | AK003086.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127221 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04714498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04714497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04231559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04877280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04714496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04685414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04634002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04298872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24129464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04314770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04685413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04282146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04103869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526795 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434214_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526792 | - | - |
| Mouse Genome 430 2.0 Array | 1434214_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434214_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66096_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66096_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500015139.mm.1 | TC0500001649.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015139.mm.1 | NM_001081108.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015139.mm.1 | NM_001081108.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015140.mm.1 | TC0500001649.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015140.mm.1 | NM_001081108.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015140.mm.1 | NM_001081108.2 | Refseq |
| Mouse Expression Set 430 | 1434214_PM_at | - | - |
| Mouse Expression Set 430 | 1434214_at | - | - |
| Murine Genome U74v2 Set | 97911_PM_at | - | - |
| Murine Genome U74v2 Set | 97911_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434214_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434214_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434214_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434214_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 912694 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7100672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621882 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621882 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621882 | - | - |

Lamtor4
Genomic Map