Assay Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026886.3 | NP_081162.1 | 10 - 11 | 1632 | 64 | |
| GenBank mRNA | AK053836.1 | - | 10 - 11 | 1364 | 64 | |
| AK129457.1 | - | 2 - 3 | 714 | 64 | ||
| BC056392.1 | - | 10 - 11 | 1622 | 6855144 | 64 | |
| JN860221.1 | - | 10 - 11 | 1279 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm4
Genomic Map