Assay Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026886.3 | NP_081162.1 | 1 - 2 | 488 | 80 | |
| GenBank mRNA | AK005085.1 | - | 1 - 2 | 488 | 80 | |
| AK053836.1 | - | 1 - 2 | 220 | 80 | ||
| AK165719.1 | - | 1 - 2 | 206 | 80 | ||
| BC056392.1 | - | 1 - 2 | 478 | 6855144 | 80 | |
| JN860221.1 | - | 1 - 2 | 135 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm4
Genomic Map