Assay Details
| Gene Symbol: Prkab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031869.2 | NP_114075.1 | 4 - 5 | 711 | 146 | |
| GenBank mRNA | AK088250.1 | - | 4 - 5 | 674 | 146 | |
| AK153552.1 | - | 4 - 5 | 731 | 146 | ||
| BC016398.1 | - | 4 - 5 | 711 | 3585002 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkab1
Genomic Map