Assay Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026805.1 | NP_081081.1 | 6 - 7 | 772 | 80 | |
| GenBank mRNA | AK029670.1 | - | 5 - 6 | 481 | 80 | |
| AK047764.1 | - | 6 - 7 | 773 | 80 | ||
| AK053833.1 | - | 6 - 7 | 763 | 80 | ||
| AK082744.1 | - | 6 - 7 | 774 | 80 | ||
| BC059878.1 | - | 6 - 7 | 725 | 6812082 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Svop
Genomic Map