Assay Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026805.1 | NP_081081.1 | 8 - 9 | 963 | 68 | |
| GenBank mRNA | AK047655.1 | - | 8 - 9 | 955 | 68 | |
| AK047764.1 | - | 8 - 9 | 964 | 68 | ||
| AK053833.1 | - | 8 - 9 | 954 | 68 | ||
| AK082744.1 | - | 8 - 9 | 965 | 68 | ||
| BC059878.1 | - | 8 - 9 | 916 | 6812082 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Svop
Genomic Map