Assay Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172541.2 | NP_766129.1 | 10 - 11 | 888 | 76 | |
| GenBank mRNA | AK028119.1 | - | 10 - 11 | 889 | 76 | |
| AK050367.1 | - | 5 - 6 | 378 | 76 | ||
| BC046757.1 | - | 10 - 11 | 848 | 5693443 | 76 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117666 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117669 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117668 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134441 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129541 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24117678 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05756313 | - | - |
| Whole Mouse Genome G4122A | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK028119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK050367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK209822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | AK211918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333349 | NM_172541.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138441 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134180 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129541 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04766942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04419398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117666 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117668 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24117669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24117665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04766945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04492800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04733555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04485782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04321874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04492791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04533837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04461995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04461997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24117650 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534518 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435031_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534504 | - | - |
| Mouse Genome 430 2.0 Array | 1435031_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435031_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215210_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215210_at | - | - |
| Mouse Expression Set 430 | 1435031_at | - | - |
| Mouse Expression Set 430 | 1435031_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435031_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435031_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381640 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3830538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3830538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003830538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2509273 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2509273 | - | - |
| mouse-6 v1.1 (Illumina) | scl25921.12.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3830538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25921.12.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2509273 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2509273 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25921.12.1_22-S | - | - |

Tmem120a
Genomic Map