Assay Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172723.4 | NP_766311.2 | 6 - 7 | 750 | 68 | |
| GenBank mRNA | AK076754.1 | - | 5 - 6 | 736 | 68 | |
| AK076768.1 | - | 6 - 7 | 507 | 68 | ||
| BC056934.1 | - | 5 - 6 | 476 | 68 | ||
| BC079634.1 | - | 5 - 6 | 544 | 68 | ||
| BC144874.1 | - | 5 - 6 | 495 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adap1
Genomic Map