Assay Details
| Gene Symbol: Srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027323.2 | NP_081599.2 | 3 - 4 | 436 | 69 | |
| GenBank mRNA | AK012640.1 | - | 3 - 4 | 401 | 69 | |
| BC030678.1 | - | 1 - 2 | 45 | 69 | ||
| BC107023.2 | - | 3 - 4 | 414 | 69 | ||
| BC125007.1 | - | 3 - 4 | 419 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrd
Genomic Map