Assay Details
| Gene Symbol: Srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027323.2 | NP_081599.2 | 4 - 5 | 530 | 60 | |
| GenBank mRNA | AK012640.1 | - | 4 - 5 | 495 | 60 | |
| BC030678.1 | - | 2 - 3 | 139 | 60 | ||
| BC125007.1 | - | 4 - 5 | 513 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991580 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121666 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | NM_027323.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | AK012640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC030678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC107023.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102002 | BC125007.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117522 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04479840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04479839 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04479846 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04479834 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04479844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04261906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04316517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04769908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04933155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04110435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04933159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04933175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04933165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23991547 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532533 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431144_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456355_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532524 | - | - |
| Mouse Genome 430 2.0 Array | 1456355_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431144_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456355_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431144_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500026753.mm.1 | TC0500003014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026753.mm.1 | NM_027323.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026753.mm.1 | NM_027323.2 | Refseq |
| Mouse Expression Set 430 | 1456355_PM_s_at | - | - |
| Mouse Expression Set 430 | 1456355_s_at | - | - |
| Mouse Expression Set 430 | 1431144_at | - | - |
| Mouse Expression Set 430 | 1431144_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456355_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456355_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456355_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456355_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431144_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431144_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 431033 | - | - |
| ABI Mouse Genome Survey Microarray | 907456 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2894057 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2894057 | - | - |

Srrd
Genomic Map