Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 4 - 5 | 505 | 93 | |
| NM_178897.4 | NP_849228.1 | 4 - 5 | 375 | 93 | ||
| GenBank mRNA | AK045279.1 | - | 4 - 5 | 376 | 93 | |
| AK082884.1 | - | 4 - 5 | 496 | 93 | ||
| AK162899.1 | - | 4 - 5 | 508 | 93 | ||
| BB619468.1 | - | 4 - 5 | 505 | 93 | ||
| BC068126.1 | - | 4 - 5 | 413 | 6829456 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyw1
Genomic Map