Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 6 - 7 | 982 | 66 | |
| NM_178897.4 | NP_849228.1 | 6 - 7 | 852 | 66 | ||
| GenBank mRNA | AK045279.1 | - | 6 - 7 | 853 | 66 | |
| AK082884.1 | - | 6 - 7 | 973 | 66 | ||
| AK162899.1 | - | 6 - 7 | 985 | 66 | ||
| BC027065.1 | - | 1 - 2 | 56 | 66 | ||
| BC068126.1 | - | 6 - 7 | 890 | 6829456 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyw1
Genomic Map