Assay Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271566.1 | NP_001258495.1 | 3 - 4 | 541 | 90 | |
| NM_001271567.1 | NP_001258496.1 | 3 - 4 | 372 | 90 | ||
| NM_001271568.1 | NP_001258497.1 | 3 - 4 | 372 | 90 | ||
| NM_009863.3 | NP_033993.2 | 3 - 4 | 372 | 90 | ||
| GenBank mRNA | AB018574.1 | - | 3 - 4 | 373 | 90 | |
| AB018575.1 | - | 3 - 4 | 373 | 90 | ||
| AB018576.1 | - | 2 - 3 | 183 | 90 | ||
| AB018577.1 | - | 2 - 3 | 183 | 90 | ||
| AJ007661.1 | - | 3 - 4 | 324 | 90 | ||
| AK012986.1 | - | 3 - 4 | 355 | 90 | ||
| AK145921.1 | - | 3 - 4 | 298 | 90 | ||
| AK146009.1 | - | 3 - 4 | 543 | 90 | ||
| AK163921.1 | - | 342 | 90 | |||
| BC062149.1 | - | 3 - 4 | 302 | 90 | ||
| BC072666.1 | - | 3 - 4 | 313 | 30611704 | 90 | |
| BC080702.1 | - | 3 - 4 | 325 | 30474846 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc7
Genomic Map