Assay Details
| Gene Symbol: Mlxip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133917.3 | NP_598678.2 | 3 - 4 | 738 | 85 | |
| NM_177582.3 | NP_808250.1 | 3 - 4 | 738 | 85 | ||
| GenBank mRNA | AK047475.1 | - | 3 - 4 | 738 | 85 | |
| AY968204.1 | - | 3 - 4 | 606 | 85 | ||
| BC137770.1 | - | 3 - 4 | 665 | 8860789 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlxip
Genomic Map