Assay Details
| Gene Symbol: Mlxip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133917.3 | NP_598678.2 | 5 - 6 | 875 | 77 | |
| NM_177582.3 | NP_808250.1 | 5 - 6 | 875 | 77 | ||
| GenBank mRNA | AK047475.1 | - | 5 - 6 | 875 | 77 | |
| AY968204.1 | - | 5 - 6 | 743 | 77 | ||
| BC137770.1 | - | 5 - 6 | 802 | 8860789 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlxip
Genomic Map