Assay Details
| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081388.1 | NP_001074857.1 | 17 - 18 | 3330 | 77 | |
| GenBank mRNA | AK038814.1 | - | 16 - 17 | 2798 | 77 | |
| AK044685.1 | - | 16 - 17 | 3430 | 77 | ||
| AK122245.2 | - | 8 - 9 | 2143 | 77 | ||
| AK147417.1 | - | 13 - 14 | 2736 | 77 | ||
| BC151100.1 | - | 13 - 14 | 2921 | 9088013 | 77 | |
| BC151101.1 | - | 13 - 14 | 2921 | 9088014 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rimbp2
Genomic Map