Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 11 - 12 | 1167 | 89 | |
| GenBank mRNA | AK049325.1 | - | 11 - 12 | 1168 | 89 | |
| AK052060.1 | - | 11 - 12 | 1155 | 89 | ||
| AK149758.1 | - | 11 - 12 | 1168 | 89 | ||
| AK165175.1 | - | 11 - 12 | 1158 | 89 | ||
| AK167405.1 | - | 11 - 12 | 1185 | 89 | ||
| AK167491.1 | - | 11 - 12 | 1185 | 89 | ||
| BC028868.1 | - | 9 - 10 | 947 | 89 | ||
| BC037595.1 | - | 11 - 12 | 1148 | 89 | ||
| BC055480.1 | - | 10 - 11 | 1069 | 89 | ||
| BC082314.1 | - | 11 - 12 | 1153 | 89 | ||
| BC106182.1 | - | 11 - 12 | 1142 | 89 | ||
| BC132172.1 | - | 11 - 12 | 1165 | 40130449 | 89 | |
| BC138093.1 | - | 11 - 12 | 1165 | 8861112 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map