Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 15 - 16 | 1745 | 104 | |
| GenBank mRNA | AK049325.1 | - | 15 - 16 | 1746 | 104 | |
| AK149758.1 | - | 15 - 16 | 1746 | 104 | ||
| AK165175.1 | - | 15 - 16 | 1736 | 104 | ||
| AK167405.1 | - | 15 - 16 | 1763 | 104 | ||
| AK167491.1 | - | 15 - 16 | 1763 | 104 | ||
| BC028868.1 | - | 13 - 14 | 1525 | 104 | ||
| BC037595.1 | - | 15 - 16 | 1726 | 104 | ||
| BC055480.1 | - | 14 - 15 | 1647 | 104 | ||
| BC082314.1 | - | 15 - 16 | 1731 | 104 | ||
| BC106182.1 | - | 15 - 16 | 1720 | 104 | ||
| BC132172.1 | - | 15 - 16 | 1743 | 40130449 | 104 | |
| BC138093.1 | - | 15 - 16 | 1743 | 8861112 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map